Assay Details
| Gene Symbol: guca1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011661.1 | NP_001011661.1 | 1 - 2 | 188 | 68 | |
| GenBank mRNA | AY850384.1 | - | 1 - 2 | 188 | 68 | |
| BC162987.1 | - | 1 - 2 | 272 | 68 | ||
| BC162992.1 | - | 1 - 2 | 272 | 68 | ||
More Information
Set Membership: |
|

guca1d
Genomic Map