Assay Details
| Gene Symbol: arf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012248.2 | NP_001012248.1 | 1 - 2 | 194 | 78 | |
| XM_005165697.4 | XP_005165754.1 | 2 - 3 | 375 | 78 | ||
| GenBank mRNA | BC163292.1 | - | 1 - 2 | 194 | 78 | |
| BC163310.1 | - | 1 - 2 | 194 | 78 | ||
More Information
Set Membership: |
|

arf3b
Genomic Map