Assay Details
| Gene Symbol: fgf11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012380.1 | NP_001012380.1 | 3 - 4 | 408 | 104 | |
| XM_021477520.1 | XP_021333195.1 | 5 - 6 | 1698 | 104 | ||
| GenBank mRNA | AB195617.1 | - | 3 - 4 | 408 | 104 | |
| BC162946.1 | - | 3 - 4 | 503 | 104 | ||
More Information
Set Membership: |
|

fgf11a
Genomic Map