Assay Details
| Gene Symbol: crygs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013276.1 | NP_001013294.1 | 2 - 3 | 318 | 85 | |
| XM_005173955.3 | XP_005174012.1 | 2 - 3 | 342 | 85 | ||
| GenBank mRNA | AY738745.1 | - | 2 - 3 | 318 | 85 | |
| BC114240.1 | - | 2 - 3 | 265 | 85 | ||
More Information
Set Membership: |
|

crygs1
Genomic Map