Assay Details
| Gene Symbol: kcnmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013525.1 | NP_001013543.1 | 4 - 5 | 566 | 67 | |
| GenBank mRNA | BC090829.1 | - | 4 - 5 | 566 | 67 | |
More Information
Set Membership: |
|

kcnmb2
Genomic Map