Assay Details
| Gene Symbol: sema3gb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014824.1 | NP_001014824.1 | 15 - 16 | 1711 | 60 | |
| XM_005167033.4 | XP_005167090.1 | 16 - 17 | 2324 | 60 | ||
| GenBank mRNA | AY766121.1 | - | 15 - 16 | 1711 | 60 | |
More Information
Set Membership: |
|

sema3gb
Genomic Map