Assay Details
| Gene Symbol: sema3gb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014824.1 | NP_001014824.1 | 5 - 6 | 525 | 62 | |
| XM_005167032.4 | XP_005167089.1 | 6 - 7 | 1138 | 62 | ||
| XM_005167033.4 | XP_005167090.1 | 6 - 7 | 1138 | 62 | ||
| GenBank mRNA | AY766121.1 | - | 5 - 6 | 525 | 62 | |
| BC163245.1 | - | 5 - 6 | 557 | 62 | ||
More Information
Set Membership: |
|

sema3gb
Genomic Map