Assay Details
| Gene Symbol: anxa1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017605.1 | NP_001017605.1 | 10 - 11 | 825 | 76 | |
| XM_021476151.1 | XP_021331826.1 | 10 - 11 | 813 | 76 | ||
| XM_021476152.1 | XP_021331827.1 | 11 - 12 | 861 | 76 | ||
| GenBank mRNA | BC092847.1 | - | 10 - 11 | 825 | 76 | |
| BC164378.1 | - | 9 - 10 | 819 | 76 | ||
More Information
Set Membership: |
|

anxa1d
Genomic Map