Assay Details
| Gene Symbol: psmd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017624.2 | NP_001017624.1 | 5 - 6 | 452 | 139 | |
| GenBank mRNA | BC092825.1 | - | 5 - 6 | 453 | 139 | |
| BC164468.1 | - | 5 - 6 | 473 | 139 | ||
More Information
Set Membership: |
|

psmd4b
Genomic Map