Assay Details
| Gene Symbol: akr1a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017640.1 | NP_001017640.1 | 6 - 7 | 827 | 65 | |
| XM_005166285.3 | XP_005166342.1 | 6 - 7 | 892 | 65 | ||
| XM_005166286.4 | XP_005166343.1 | 6 - 7 | 850 | 65 | ||
| XM_005166287.3 | XP_005166344.1 | 6 - 7 | 961 | 65 | ||
| XM_005166289.4 | XP_005166346.1 | 6 - 7 | 941 | 65 | ||
| GenBank mRNA | BC092808.1 | - | 6 - 7 | 827 | 65 | |
| BC155150.1 | - | 6 - 7 | 806 | 65 | ||
More Information
Set Membership: |
|

akr1a1b
Genomic Map