Assay Details
| Gene Symbol: ppp1r14aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017863.1 | NP_001017863.1 | 3 - 4 | 336 | 84 | |
| GenBank mRNA | BC092987.1 | - | 3 - 4 | 336 | 84 | |
More Information
Set Membership: |
|

ppp1r14aa
Genomic Map