Assay Details
| Gene Symbol: mul1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018124.1 | NP_001018134.1 | 3 - 4 | 264 | 68 | |
| GenBank mRNA | BC088385.1 | - | 3 - 4 | 264 | 68 | |
More Information
Set Membership: |
|

mul1a
Genomic Map