Assay Details
| Gene Symbol: nip7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020590.1 | NP_001018426.1 | 3 - 4 | 368 | 56 | |
| GenBank mRNA | BC095238.1 | - | 3 - 4 | 368 | 56 | |
| BC164958.1 | - | 3 - 4 | 320 | 56 | ||
More Information
Set Membership: |
|

nip7
Genomic Map