Assay Details
| Gene Symbol: ints11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020621.1 | NP_001018457.1 | 14 - 15 | 1510 | 106 | |
| XM_005166209.4 | XP_005166266.1 | 13 - 14 | 1681 | 106 | ||
| GenBank mRNA | BC095364.1 | - | 14 - 15 | 1510 | 106 | |
More Information
Set Membership: |
|

ints11
Genomic Map