Assay Details
| Gene Symbol: ints11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020621.1 | NP_001018457.1 | 3 - 4 | 231 | 73 | |
| XM_005166209.4 | XP_005166266.1 | 2 - 3 | 402 | 73 | ||
| GenBank mRNA | BC095364.1 | - | 3 - 4 | 231 | 73 | |
More Information
Set Membership: |
|

ints11
Genomic Map