Assay Details
| Gene Symbol: ap5m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020654.1 | NP_001018490.1 | 1 - 2 | 271 | 65 | |
| XM_005158894.3 | XP_005158951.1 | 1 - 2 | 288 | 65 | ||
| GenBank mRNA | BC095676.1 | - | 1 - 2 | 271 | 65 | |
More Information
Set Membership: |
|

ap5m1
Genomic Map