Assay Details
| Gene Symbol: isca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025178.2 | NP_001020349.1 | 2 - 3 | 186 | 85 | |
| GenBank mRNA | BC097234.1 | - | 2 - 3 | 187 | 85 | |
More Information
Set Membership: |
|

isca1
Genomic Map