Assay Details
| Gene Symbol: lrrc8aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029949.1 | NP_001025120.1 | 6 - 7 | 2214 | 82 | |
| GenBank mRNA | BC097172.1 | - | 6 - 7 | 2214 | 82 | |
More Information
Set Membership: |
|

lrrc8aa
Genomic Map