Assay Details
| Gene Symbol: gpc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037373.1 | NP_001032450.1 | 2 - 3 | 723 | 83 | |
| GenBank mRNA | BC092886.1 | - | 2 - 3 | 646 | 83 | |
| BC109411.1 | - | 2 - 3 | 723 | 83 | ||
More Information
Set Membership: |
|

gpc1a
Genomic Map