Assay Details
| Gene Symbol: gpc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037373.1 | NP_001032450.1 | 5 - 6 | 1416 | 88 | |
| GenBank mRNA | BC092886.1 | - | 5 - 6 | 1339 | 88 | |
| BC109411.1 | - | 5 - 6 | 1416 | 88 | ||
| KC836776.1 | - | 5 - 6 | 1009 | 88 | ||
More Information
Set Membership: |
|

gpc1a
Genomic Map