Assay Details
| Gene Symbol: gpc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037373.1 | NP_001032450.1 | 6 - 7 | 1553 | 78 | |
| GenBank mRNA | BC092886.1 | - | 6 - 7 | 1476 | 78 | |
| BC109411.1 | - | 6 - 7 | 1553 | 78 | ||
| KC836776.1 | - | 6 - 7 | 1146 | 78 | ||
More Information
Set Membership: |
|

gpc1a
Genomic Map