Assay Details
| Gene Symbol: bpnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037568.1 | NP_001032657.1 | 1 - 2 | 672 | 71 | |
| GenBank mRNA | BC110106.1 | - | 1 - 2 | 672 | 71 | |
More Information
Set Membership: |
|

bpnt2
Genomic Map