Assay Details
| Gene Symbol: slc6a6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037661.1 | NP_001032750.1 | 1601 | 70 | ||
| GenBank mRNA | AY842304.1 | - | 1601 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

slc6a6b| Gene Symbol: slc6a6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037661.1 | NP_001032750.1 | 1601 | 70 | ||
| GenBank mRNA | AY842304.1 | - | 1601 | 70 | ||
Set Membership: |
|