Assay Details
| Gene Symbol: scn4aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039825.1 | NP_001034914.1 | 19 - 20 | 3339 | 95 | |
| XM_017358479.2 | XP_017213968.1 | 20 - 21 | 3525 | 95 | ||
| GenBank mRNA | DQ149506.1 | - | 19 - 20 | 3336 | 95 | |
| DQ351532.1 | - | 8 - 9 | 1527 | 95 | ||
More Information
Set Membership: |
|

scn4aa
Genomic Map