Assay Details
| Gene Symbol: scn4aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039825.1 | NP_001034914.1 | 20 - 21 | 3622 | 70 | |
| XM_017358479.2 | XP_017213968.1 | 21 - 22 | 3808 | 70 | ||
| GenBank mRNA | DQ149506.1 | - | 20 - 21 | 3619 | 70 | |
| DQ351532.1 | - | 9 - 10 | 1810 | 70 | ||
More Information
Set Membership: |
|

scn4aa
Genomic Map