Assay Details
| Gene Symbol: scn4aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039825.1 | NP_001034914.1 | 23 - 24 | 3909 | 67 | |
| XM_017358479.2 | XP_017213968.1 | 24 - 25 | 4095 | 67 | ||
| GenBank mRNA | DQ351532.1 | - | 12 - 13 | 2097 | 67 | |
More Information
Set Membership: |
|

scn4aa
Genomic Map