Assay Details
| Gene Symbol: scn4aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039825.1 | NP_001034914.1 | 6 - 7 | 936 | 103 | |
| XM_017358479.2 | XP_017213968.1 | 7 - 8 | 103 | |||
| GenBank mRNA | DQ149506.1 | - | 6 - 7 | 103 | ||
More Information
Set Membership: |
|

scn4aa
Genomic Map