Assay Details
| Gene Symbol: selenoo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044871.3 | NP_001038336.2 | 1 - 2 | 935 | 64 | |
| GenBank mRNA | EH495893.1 | - | 1 - 2 | 675 | 64 | |
More Information
Set Membership: |
|

selenoo1
Genomic Map