Assay Details
| Gene Symbol: selenoo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044871.3 | NP_001038336.2 | 3 - 4 | 1320 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

selenoo1| Gene Symbol: selenoo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044871.3 | NP_001038336.2 | 3 - 4 | 1320 | 81 | |
Set Membership: |
|