Assay Details
| Gene Symbol: selenoo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044871.3 | NP_001038336.2 | 7 - 8 | 2082 | 62 | |
| GenBank mRNA | EE689105.1 | - | 3 - 4 | 418 | 62 | |
More Information
Set Membership: |
|

selenoo1
Genomic Map