Assay Details
| Gene Symbol: erf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044927.1 | NP_001038392.1 | 3 - 4 | 530 | 68 | |
| XM_005159364.4 | XP_005159421.1 | 3 - 4 | 901 | 68 | ||
| GenBank mRNA | BC171656.1 | - | 3 - 4 | 471 | 68 | |
More Information
Set Membership: |
|

erf
Genomic Map