Assay Details
| Gene Symbol: kcnh1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044931.1 | NP_001038396.1 | 9 - 10 | 1825 | 74 | |
| XM_009293096.3 | XP_009291371.1 | 11 - 12 | 2031 | 74 | ||
| XM_009293097.2 | XP_009291372.1 | 11 - 12 | 2031 | 74 | ||
| XM_009293098.3 | XP_009291373.1 | 10 - 11 | 2004 | 74 | ||
| XM_009293099.3 | XP_009291374.1 | 10 - 11 | 2004 | 74 | ||
| GenBank mRNA | BC162812.1 | - | 9 - 10 | 1865 | 74 | |
| DQ022612.1 | - | 9 - 10 | 1825 | 74 | ||
| HQ703597.1 | - | 9 - 10 | 1825 | 74 | ||
More Information
Set Membership: |
|

kcnh1a
Genomic Map