Assay Details
| Gene Symbol: nfatc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045159.1 | NP_001038624.1 | 8 - 9 | 2235 | 67 | |
| GenBank mRNA | DQ530435.1 | - | 8 - 9 | 2235 | 67 | |
More Information
Set Membership: |
|

nfatc1
Genomic Map