Assay Details
| Gene Symbol: asap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045207.1 | NP_001038672.1 | 10 - 11 | 1375 | 61 | |
| XM_005158734.4 | XP_005158791.1 | 11 - 12 | 1405 | 61 | ||
| XM_021467217.1 | XP_021322892.1 | 12 - 13 | 1432 | 61 | ||
| XM_021467218.1 | XP_021322893.1 | 11 - 12 | 1423 | 61 | ||
| XM_021467219.1 | XP_021322894.1 | 12 - 13 | 1432 | 61 | ||
More Information
Set Membership: |
|

asap2a
Genomic Map