Assay Details
| Gene Symbol: asap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045207.1 | NP_001038672.1 | 12 - 13 | 1529 | 75 | |
| XM_005158734.4 | XP_005158791.1 | 13 - 14 | 1559 | 75 | ||
| XM_021467217.1 | XP_021322892.1 | 14 - 15 | 1586 | 75 | ||
| XM_021467218.1 | XP_021322893.1 | 13 - 14 | 1577 | 75 | ||
| XM_021467219.1 | XP_021322894.1 | 14 - 15 | 1586 | 75 | ||
More Information
Set Membership: |
|

asap2a
Genomic Map