Assay Details
| Gene Symbol: asap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045207.1 | NP_001038672.1 | 18 - 19 | 2247 | 71 | |
| XM_005158734.4 | XP_005158791.1 | 19 - 20 | 2277 | 71 | ||
| XM_021467217.1 | XP_021322892.1 | 20 - 21 | 2304 | 71 | ||
| XM_021467218.1 | XP_021322893.1 | 19 - 20 | 2295 | 71 | ||
| XM_021467219.1 | XP_021322894.1 | 20 - 21 | 2304 | 71 | ||
More Information
Set Membership: |
|

asap2a
Genomic Map