Assay Details
| Gene Symbol: asap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045207.1 | NP_001038672.1 | 27 - 28 | 3315 | 100 | |
| XM_005158734.4 | XP_005158791.1 | 28 - 29 | 3345 | 100 | ||
| XM_021467217.1 | XP_021322892.1 | 29 - 30 | 3372 | 100 | ||
| XM_021467218.1 | XP_021322893.1 | 28 - 29 | 3363 | 100 | ||
| XM_021467219.1 | XP_021322894.1 | 29 - 30 | 3345 | 100 | ||
More Information
Set Membership: |
|

asap2a
Genomic Map