Assay Details
| Gene Symbol: washc2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003199716.5 | XP_003199764.1 | 4 - 5 | 556 | 83 | |
| XM_005156704.4 | XP_005156761.1 | 4 - 5 | 556 | 83 | ||
| XM_005156705.4 | XP_005156762.1 | 4 - 5 | 556 | 83 | ||
| XM_009307266.3 | XP_009305541.1 | 4 - 5 | 543 | 83 | ||
| XM_017358961.2 | XP_017214450.1 | 4 - 5 | 556 | 83 | ||
| XM_021480773.1 | XP_021336448.1 | 4 - 5 | 556 | 83 | ||
| GenBank mRNA | BC122360.1 | - | 4 - 5 | 548 | 83 | |
| BC171336.1 | - | 4 - 5 | 495 | 83 | ||
| BC171338.1 | - | 4 - 5 | 495 | 83 | ||
More Information
Set Membership: |
|

washc2c
Genomic Map