Assay Details
| Gene Symbol: itln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076622.2 | NP_001070090.1 | 2 - 3 | 99 | 90 | |
| GenBank mRNA | BC124294.1 | - | 2 - 3 | 102 | 90 | |
| BC164264.1 | - | 1 - 2 | 88 | 90 | ||
| EU583680.1 | - | 1 - 2 | 82 | 90 | ||
More Information
Set Membership: |
|

itln1
Genomic Map