Assay Details
| Gene Symbol: itln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076622.2 | NP_001070090.1 | 6 - 7 | 734 | 105 | |
| XM_017355768.2 | XP_017211257.1 | 6 - 7 | 703 | 105 | ||
| GenBank mRNA | BC124294.1 | - | 6 - 7 | 737 | 105 | |
| BC164264.1 | - | 5 - 6 | 723 | 105 | ||
| EU583680.1 | - | 5 - 6 | 717 | 105 | ||
More Information
Set Membership: |
|

itln1
Genomic Map