Assay Details
| Gene Symbol: itln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076622.2 | NP_001070090.1 | 7 - 8 | 838 | 133 | |
| XM_017355768.2 | XP_017211257.1 | 7 - 8 | 807 | 133 | ||
| GenBank mRNA | BC124294.1 | - | 7 - 8 | 841 | 133 | |
| BC164264.1 | - | 6 - 7 | 827 | 133 | ||
| EU583680.1 | - | 6 - 7 | 821 | 133 | ||
More Information
Set Membership: |
|

itln1
Genomic Map