Assay Details
| Gene Symbol: abi1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076707.1 | NP_001070175.1 | 4 - 5 | 659 | 108 | |
| XM_005171176.4 | XP_005171233.1 | 5 - 6 | 699 | 108 | ||
| XM_005171177.4 | XP_005171234.1 | 5 - 6 | 700 | 108 | ||
| GenBank mRNA | BC124378.1 | - | 4 - 5 | 659 | 108 | |
More Information
Set Membership: |
|

abi1b
Genomic Map