Assay Details
| Gene Symbol: slc27a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077248.1 | NP_001070716.1 | 2 - 3 | 377 | 60 | |
| XM_005160200.4 | XP_005160257.1 | 2 - 3 | 414 | 60 | ||
| GenBank mRNA | BC076535.1 | - | 1 - 2 | 120 | 60 | |
| BC109455.1 | - | 2 - 3 | 217 | 60 | ||
| BC124752.1 | - | 2 - 3 | 377 | 60 | ||
| BC164558.1 | - | 1 - 2 | 212 | 60 | ||
More Information
Set Membership: |
|

slc27a1b
Genomic Map