Assay Details
| Gene Symbol: slc27a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077248.1 | NP_001070716.1 | 9 - 10 | 1536 | 114 | |
| XM_005160200.4 | XP_005160257.1 | 9 - 10 | 1573 | 114 | ||
| GenBank mRNA | BC076535.1 | - | 8 - 9 | 1279 | 114 | |
| BC124752.1 | - | 9 - 10 | 1536 | 114 | ||
| BC164558.1 | - | 8 - 9 | 1371 | 114 | ||
More Information
Set Membership: |
|

slc27a1b
Genomic Map