Assay Details
| Gene Symbol: prkar2aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077370.2 | NP_001070838.2 | 6 - 7 | 1189 | 64 | |
| GenBank mRNA | BC124725.1 | - | 6 - 7 | 1190 | 64 | |
| BC164493.1 | - | 6 - 7 | 829 | 64 | ||
| BQ481277.1 | - | 3 - 4 | 305 | 64 | ||
More Information
Set Membership: |
|

prkar2aa
Genomic Map