Assay Details
| Gene Symbol: usp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077449.1 | NP_001070917.1 | 34 - 35 | 5517 | 79 | |
| GenBank mRNA | DQ086492.1 | - | 34 - 35 | 5517 | 79 | |
More Information
Set Membership: |
|

usp9
Genomic Map