Assay Details
| Gene Symbol: usp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077449.1 | NP_001070917.1 | 41 - 42 | 7048 | 91 | |
| XM_005167736.4 | XP_005167793.1 | 42 - 43 | 7548 | 91 | ||
| XM_009304755.3 | XP_009303030.1 | 42 - 43 | 7549 | 91 | ||
| XM_009304756.3 | XP_009303031.1 | 42 - 43 | 7549 | 91 | ||
| XM_009304757.3 | XP_009303032.1 | 42 - 43 | 7550 | 91 | ||
| GenBank mRNA | DQ086492.1 | - | 41 - 42 | 7048 | 91 | |
More Information
Set Membership: |
|

usp9
Genomic Map