Assay Details
| Gene Symbol: irg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077607.1 | NP_001071075.1 | 5 - 6 | 664 | 69 | |
| GenBank mRNA | BC125936.1 | - | 5 - 6 | 664 | 69 | |
More Information
Set Membership: |
|

irg1l
Genomic Map