Assay Details
| Gene Symbol: mtus1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077608.2 | NP_001071076.2 | 7 - 8 | 1023 | 83 | |
| XM_005157240.4 | XP_005157297.1 | 12 - 13 | 3311 | 83 | ||
| XM_005157242.4 | XP_005157299.1 | 6 - 7 | 718 | 83 | ||
| XM_017359019.2 | XP_017214508.1 | 12 - 13 | 3308 | 83 | ||
| XM_021481039.1 | XP_021336714.1 | 9 - 10 | 1258 | 83 | ||
| GenBank mRNA | BC125970.1 | - | 7 - 8 | 1023 | 83 | |
More Information
Set Membership: |
|

mtus1a
Genomic Map