Assay Details
| Gene Symbol: mtus1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077608.2 | NP_001071076.2 | 8 - 9 | 1131 | 85 | |
| XM_005157240.4 | XP_005157297.1 | 13 - 14 | 3419 | 85 | ||
| XM_005157242.4 | XP_005157299.1 | 7 - 8 | 826 | 85 | ||
| XM_017359019.2 | XP_017214508.1 | 13 - 14 | 3416 | 85 | ||
| XM_021481039.1 | XP_021336714.1 | 10 - 11 | 1366 | 85 | ||
| GenBank mRNA | BC125970.1 | - | 8 - 9 | 1131 | 85 | |
More Information
Set Membership: |
|

mtus1a
Genomic Map